Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-450 Epoxy

Catalog No. 14011
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-011 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-011 Supplier Invitrogen™ Supplier No. 14011
Add to Cart
Edge
Add to Cart

Dynabeads™ M-450 Epoxy beads are ideal for custom cell-isolation protocols upstream of molecular applications.

Dynabeads™ M-450 Epoxy beads are ideal for custom cell-isolation protocols upstream of molecular applications. Purified antibodies can be used in conjunction with these beads to create a magnetic cell separation media with the good antibody orientation.

Ideal for Cell Separation when you have a custom antibody:
Low background and covalent linking of antibodies to the bead surface make these beads an excellent choice for cell separation applications when a downstream release from the beads is not required.

Ultra rapid protocols:
• Quickly isolate targeted cell populations upstream of molecular analyses
• Excellent for cell isolation procedures that require custom antibodies
• Couple different custom antibodies to different beads to select specific subpopulations of cells

Not recommended for Immunoprecipitation (IP):
These 4.5 micron beads are not recommended for IP as they have a lower surface capacity per unit mass when compared to 1 μm and 2.8 μm Tosylactivated and Epoxy coated Dynabeads™. Use rather Dynabeads Protein A (10001D, 10002D), Dynabeads Protein G (10003D, 10004D) or one of the Dynabeads Immunoprecipitation Kits (10006D, 10007D)

Coupling procedure outline:
Any purified antibody can be covalently coupled to the bead surface in a easy-to-perform overnight reaction. Simply incubate the antibody you wish to use for your cell isolation procedure with these 4.5 μm Dynabeads™. Optimal coupling occurs at high pH (8.5-9.5) and at 37°C. For pH labile antibodies, coupling can be performed in an alternative buffer at pH 7.4.

About Dynabeads™:
Dynabeads™ are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling antibodies coupled to the beads to continuously interact with the cell suspension. These 4.5 μm superparamagnetic beads experience a strong pull when placed in a magnetic field and gently pull targeted cells to the tube walls when the tube is transferred to a rack containing a strong magnetic field. With the beads and attached cells concentrated at the tube wall the supernatant liquid containing non-targeted cells is easily and quickly decanted or pipetted away. Washing steps are performed similarly.

Bead Characteristics for Dynabeads™ M-450 Epoxy:
• Coated in glycidyl ether (Epoxy) groups
• Hydrophobic, pH Neutral
• Covalent binding by primary amine (NH2) or sulphydryl (SH) groups
• Large bead 4.5 μm bead size is ideal for cellular isolations
• Large bead 4.5 μm bead size not recommended for IP applications

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type All cells from all species
Content And Storage Store at 2°C to 8°C.
Product Type Epoxy Bead
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 cells per isolation
Target Species All species
Diameter (Metric) 4.5 μm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.