Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-450 Tosylactivated

Catalog No. 14013
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-013 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-013 Supplier Invitrogen™ Supplier No. 14013
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

5mL beads at a concentration of 4 x 108 beads/mL in distilled water

Dynabeads™ M-450 Tosylactivated are 4.5 μm superparamagnetic beads containing surface tosyl groups.

Dynabeads™ M-450 Tosylactivated are 4.5 μm superparamagnetic beads containing surface tosyl groups. The beads covalently bind primary amino and sulfhydryl groups in antibodies to position them in the optimal orientation for cell-surface protein binding. Low background and covalent binding of the antibodies make these beads an excellent choice for cell separation applications when a downstream release from the beads is not required.

• Quickly deplete cells or positively isolate target cells for downstream molecular analysis
• Excellent for cell isolation procedures using your own unique antibody
• Several antibodies can be coupled on the same beads for non-cell-based applications, such as cell activation

Coupling procedure
Covalent coupling is performed overnight by incubating the antibody of choice with the Dynabeads™ M-450 Tosylactivated beads. Optimal coupling occurs at high pH (8.5–9.5) and at 37°C. For pH-labile antibodies, coupling can be performed in an alternative buffer at pH 7.4.

Not recommended for immunoprecipitation (IP)
These 4.5 μm Tosylactivated beads are not recommended for IP as they have a lower surface capacity per unit mass when compared to 1 μm and 2.8 μm Tosylactivated and Epoxy Dynabeads™.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type All cells from all species
Content And Storage Contains: 5 mL beads at a concentration of 4 x 108 beads/mL in distilled water.
Storage: 2°C to 8°C.
Product Type Tosylactivated Bead
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 cells per isolation
Target Species All species
Diameter (Metric) 4.5 μm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.