Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ MyOne™ Tosylactivated

Catalog No. p-4924213
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-501 2 mL
65-502 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-501 Supplier Invitrogen™ Supplier No. 65501
Only null left
Add to Cart
Edge
Add to Cart

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated.

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated. Optimal antibody orientation makes these beads an excellent choice for the immunoprecipitation of proteins and protein complexes.

Ideal for Immunoprecipitation of protein complexes:
Low background and covalent linking of antibodies to the bead surface make Dynabeads Tosylactivated an excellent choice for Immunoprecipitation of proteins and protein complexes (Co-Immunoprecipitation, Co-IP). Gentle and rapid magnetic concentration of the beads and short incubation times (possible due to fast surface-based binding kinetics) make Dynabeads Tosylactivated an excellent choice for the immunoprecipitation of highly-labile or and/or transient (short-lived) protein complexes.

Bead Surface Characteristics for Dynabeads Tosylactivated:
• p-toluene-sulfonyl (Tosyl) groups
• Hydrophobic, pH Neutral
• Covalent binding by primary amine (NH2) or sulphydryl (SH) groups

Ultra rapid protocols:
• Identify members of protein complexes in minutes rather than hours
• Temporal resolution short enough to identify transient and labile complexes
• Identify binding partners that cannot be identified with longer protocols
• Rapid protocols further reduce the already ultra-low background binding
• Stronger signal to noise-ratios

Applications:
• IP of proteins and protein complexes
• Couple functional enzymes to bead surface for downstream assays
• Couple peptides to bead surface to identify binders

Purify peptides, proteins and enzymes that are:
• Transiently stable
• Structurally intact
• Temperature labile
• In their native conformation and functional

Coupling procedure outline:
Covalent coupling is performed overnight by incubating the desired ligand with the Dynabeads Tosylactivated. Ligands commonly coupled to Dynabeads Tosylactivated include peptides and proteins (e.g. antibodies for Immunoprecipitation or Co-Immunoprecipitation). Coupling occurs at neutral to high pH and at 37°C. We recommend coupling at pH 8.5-9.5, but for pH labile ligands, coupling can be performed in an alternative buffer at pH 7.4.

Upon completion of the ligand coupling step, the actual Dynabeads Tosylactivated surface coating will be rendered inert, resulting in low non-specific binding.

Binding capacity per milligram beads:
Varies depending on ligand, (e.g. 14-19 μg IgG)

About Dynabeads:

Dynabeads are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling ligands coupled to the beads to continuously interact with the entire sample volume. The superparamagnetic beads are pulled to the tube walls by transferring the tube to a rack containing a strong magnetic field. Strong magnetic fields quickly pull the beads to the tube wall, allowing for easy and complete removal of the supernatant by pipette. Washing steps are performed similarly.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C.
Elution Volume Min 5 μL
High-throughput Compatibility High-throughput Compatible
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality p-Toluene-Sulfonyl, Tosylactivated
Product Line DYNAL, Dynabeads, MyOne
Quantity 2 mL
Type Tosylactivated Bead
Format Beads in solution
Purity or Quality Grade Research Grade
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.