Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Sample Loading Buffer, 1X
GREENER_CHOICE

Catalog No. 10482055
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 1.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-482-055 4 x 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-482-055 Supplier Invitrogen™ Supplier No. 10482055
  • $85.00
    $81.65 / Each of 1

    Save $3.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

E-Gel™ Sample Loading Buffer is supplied as a 1X solution in buffer containing 0.0094% XCFF and 0.06% tartrazine

Supplied as a ready-to-use 1X solution

E-Gel™ Sample Buffer is formulated specifically for maximum performance on E-Gel EX gels as well as other E-Gel precast gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 4 x 1.25 mL 1X E-Gel Sample Loading buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

Buffer Sample Loading Buffers
Concentration 1X
Gel Type Agarose
Label or Dye Tartrazine, Xylene Cyanol, Xylene Cyanol FF
Sample Loading Volume 1.25 mL
Form Liquid
Gel Compatibility E-Gel™ agarose gels
Quantity 4 x 1.25 mL
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Product Line E-Gel
Product Type Sample Loading Buffer
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.