Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ElectroMAX™ A. tumefaciens LBA4404 Cells

Catalog No. 18313015
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 40 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-313-015 5 x 40 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-313-015 Supplier Invitrogen™ Supplier No. 18313015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agrobacterium cells that have been specially prepared for transformation by electroporation

  • Contain disarmed Ti plasmid pAL 4404, which has only vir and ori region of Ti plasmid
  • Used to transform variety of plant species, including tobacco and Arabidopsis, at high efficiency
  • Transformation efficiency: > 5x106
  • Electrocompetent

Not high throughput compatible (manual)

Cloning, Transformation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Format Tube(s)
Product Type Tumefaciens Cell
Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Bacterial or Yeast Strain A. tumefaciens LBA4404
Blue/White Screening No
High-throughput Compatibility Not High-throughput Compatible (Manual)
Species A. tumefaciens
Shipping Condition Dry Ice
Quantity 5 x 40 μL
Product Line ElectroMAX

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.