Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EveryPrep™ Universal Vacuum Manifold

Catalog No. K211101
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K211101 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K211101 Supplier Invitrogen™ Supplier No. K211101
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids.

The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids. It offers the ability to directly elute from the manifold using the new ChargeSwitch™ Pro Filter Plasmid Mini, Midi and Maxi Kits and also many other competitors' vacuum assisted spin column preps and filter plates. The EveryPrep™ Universal Vacuum Manifold replaces and consolidates all previous manifolds into one space saving benchtop unit. The manifold reduces sample handling to a minimum by allowing direct parallel and simultaneous processing of up to 96 samples using collection plates, deep-well blocks, or microtubes. The use of a vacuum eliminates the need for time-consuming centrifugation. The manifold will consist of a dual chamber construct fashioned out of aluminum with interchangeable top plates of a luer-lock tapped top plate, a 96-well top plate, a mini column elution plate and a maxi column elution plate.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Vacuum Manifold
Content And Storage 1 vacuum manifold. Store at room temperature.
Quantity 1 each
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.