Learn More
Invitrogen™ pEXP1-DEST Vector Kit
Catalog No. V96001
Encompass_Preferred
Change view
Click to view available options
Quantity:
6 μg
Catalog No. Quantity
V96001 6 μg
1 options
Catalog No. V96001 Supplier Invitrogen™ Supplier No. V96001
Add to Cart
Add to Cart
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase enzyme mix, and reaction buffer.
Destination vectors have attR sites for recombination with any attL-flanked fragment
Cell Free Protein Expression for Functional Analysis, Cell-Free (in vitro) Expression, Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7
Product Type Gateway System Destination Expression Vector
Selection Agent (Eukaryotic) None
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cleavage EK (Enterokinase) Recognition Site
Protein Tag His Tag (6x)
Cloning Method Gateway
Quantity 6 μg
Vector pEXP, pDEST
Product Line Expressway, Gateway
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.