Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Flp-In™-CHO Cell Line

Catalog No. R75807
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R75807 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R75807 Supplier Invitrogen™ Supplier No. R75807
  • $2,840.00
    $2,612.65 / Each of 1

    Save $227.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

3 x 106 cells are supplied frozen in 1mL 90% complete medium and 10% DMSO.

Designed for rapid generation of stable cell lines that express protein of interest from Flp-In expression vector

  • These cells contain single stably integrated FRT site at transcriptionally active genomic locus
  • Targeted integration of Flp-Inexpression vector ensures high-level expression of gene of interest
  • Created by transfecting parent cell lines with pFRT/lacZeo and selecting for stable Zeocin™-resistant clones

  • The Flp-In-CHO Cell Line was created by transfecting CHO cells with pFRT/lacZeo2 and selecting for Zeocin™-resistant clones
  • Flp-In-CHO cell lines work well with Flp-In vectors that express gene from CMV promoter (pcDNA™5/FRT, pcDNA5/FRT/V5-His-TOPO™, and pSecTag/FRT/V5-His-TOPO™)

Constitutive Expression, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 3 x 106 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.
Cell Line Flp-In™-CHO Cell Line
Species Hamster
Protein Tag pSec
Quantity 1 mL
Product Line Flp-In
Is multiple integration of the Flp-In expression construct possible? How do you screen for multiple integrants, and how stable is the Flp-In expression cell line?

In theory, one can get multiple integrations of the Flp-In expression construct—an FRT-specific integration event and a random, second-site integration. However, random integration is a relatively uncommon event. Limiting the amount of DNA in the transfection will reduce the chance of second-site integration. We have transfected 293 cells (lacking the FRT site) with the pcDNA5/FRT vector and have identified one potential second-site integrant after screening over 200 clones. DNA integrations can be detected by Southern blot. A single integrant will display a single band; double: two; triple: three, etc. We have maintained a number of Flp-In expression cell lines for over four months and have not observed any loss of the Flp-In expression construct, whether hygromycin selection was maintained or not.

What kind of Flp-In T-REx cell lines do you offer?

We offer the Flp-In T-REx system that contains pFRT/lacZeo and pcDNA6/TR stably integrated into HEK 293 cells. This cell line has been functionally tested for its ability to regulate expression.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.