Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTP

Catalog No. 18332015
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-332-015 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-332-015 Supplier Invitrogen™ Supplier No. 18332015
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATP, CTP, GTP, UTP (nucleoside 5'-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts.

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).

Related products
• 10 mM ATP (Cat. No. 18330019)
• 10 mM CTP (18331017)
• 10 mM UTP (18333013)
• 10 mM NTP Mix (18109017)

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10 mM GTP in Tris-HCl (250 μL)

Store at –5 to –30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Concentration 10 mM
Label or Dye Unlabeled
Quantity 500 μL
For Use With (Application) in vitro Transcription
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.