Learn More
Invitrogen™ InsectSelect™ Glow Kit with Sf9 Cells
Catalog No. K81001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
K81001 1 kit
1 options
Catalog No. K81001 Supplier Invitrogen™ Supplier No. K81001
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart
Includes
The InsectSelect Glow Kit includes 20μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1g Zeocin, Cellfectin Reagent, Sf9 cells, and 1L of GIBCO Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described.
Features the pIZT/V5-His vector for high-level expression of the gene of interest
  • The pIZT/V5-His vector features the OpIE2 promoter for constitutive expression
  • The Zeocin resistance gene for rapid selection of stably transfected cell lines
  • C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Anti-V5 Antibody and rapid purification with nickel-chelating resin
  • pIZT/V5-His expresses a fusion of the green fluorescent protein and the Zeocin resistanc protein (Zeo-GFP): The Zeo-GFP fusion protein permits rapid selection of stably transfected cell lines with Zeocin and confers a fluorescent phenotype that simplifies identification of transfected cells
Constitutive Protein Expression in Insect Cells, High-level Protein Production in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter OplE2
Product Type Sf9 Cells Glow Kit
Content And Storage The InsectSelect™ Glow Kit includes 20 μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2 μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1 g Zeocin,™ Cellfectin™ Reagent, Sf9 cells, and 1 L of GIBCO™ Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above. Store frozen insect cells in liquid nitrogen. Store medium and Cellfectin™ Reagent at +4°C. Store all other reagents at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag Position (to your gene) C-terminal
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pIZ
Product Line InsectSelect
Does the incubator need to be humidified when culturing insect cells?
Humidification is only necessary for monolayer cultures and suspension cultures with volumes less than 35 mL. With shaker or spinner cultures greater than 35 mL, humidification is not necessary.
For Research Use Only. Not for use in diagnostic procedures.