Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Low DNA Mass Ladder
GREENER_CHOICE

Catalog No. 10068013
Encompass_Preferred
Change view
Click to view available options
Quantity:
23.5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-068-013 23.5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not heat the ladder before loading.
Catalog No. 10-068-013 Supplier Invitrogen™ Supplier No. 10068013
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Do not heat the ladder before loading.

Includes

Low DNAmass™ Ladder, 200μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA

Suitable for estimating the mass (quantity) of unknown DNA samples by ethidium bromide staining

Invitrogen Low DNA Mass Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. Low DNA Mass Ladder consist of six individual chromatography-purified DNA fragments.

Low DNA Mass Ladder is ideal for separation on 1–2% agarose gels.

Highlights of Low DNA Mass Ladder
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
Precise—an exact amount of DNA in each band

Product Use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with DNA bands of increasing intensity. An exact amount of DNA in each band allows approximate quantification of DNA samples.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 200 µL Low DNA Mass Ladder
• 1 mL10X BlueJuice Gel Loading Buffer

Store at -20°C.

Concentration 0.1175 μg/μL
Gel Type Agarose
Ready to Load No
Sample Loading Volume 1 mL
Size Range 100 to 2000 bp
Volume (Metric) 200 μL
Gel Compatibility Agarose gel
No. of Reactions 50 Applications
Quantity 23.5 μg
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.