Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pAc5.1/V5-His A, B, & C Vectors

Catalog No. V411020
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V411020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V411020 Supplier Invitrogen™ Supplier No. V411020
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains 20μg each of a selection vector (pCoBlast or pCoHygro), pAc5.1/V5-His A, B, C, and pAc5.1/V5-His/lacZ.

For high-level constitutive expression of recombinant proteins

  • The vector offers the strong, constitutive promoter from the Drosophila actin 5C gene
  • Small size (5.4 kb) to improve DNA yields and increase subcloning efficiency
  • C-terminal V5 epitope tag for rapid detection with Anti-V5 Antibody
  • C-terminal 6xHis tag for simple purification of recombinant fusion proteins using nickel-chelating resin
  • A set of three vectors-A, B, and C-is provided: Each vector has the multiple cloning site in a different reading frame relative to the coding sequence of the C-terminal tag

Constitutive Protein Expression in Insect Cells, High-level Protein Production in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Format Lyophilized
Promoter pAC5
Product Type Drosophila Expression Vector
Content And Storage Contains 20 µg each of lyophilized pAc5.1/V5-His A, B, C and pAc5.1/V5-His/LacZ.
Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 20 μg
Vector pAC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.