Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pcDNA™3.1 Directional TOPO™ Expression Kit

Catalog No. p-4925865
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K490001 20 Reactions
K490040 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K490001 Supplier Invitrogen™ Supplier No. K490001
Only null left
Add to Cart
Add to Cart

Includes

Box 1: 20μL pcDNA 3.1D/V5-His-TOPO (15–20ng/μL), 10μL dNTP Mix, 50μL Salt Solution, 1mL Sterile Water, 20μL T7 Sequencing Primer (0.1μg/μL), 20μL BGH Reverse Sequencing Primer (0.1μg/μL), 10μL Control PCR Template (0.1μg/μL), 10μL Control PCR Primers (0.1μg/μL each, 10μL Expression Plasmid (0.5μg/μL); Box 2: 6mL S.O.C. Medium, 21 × 50μL TOP10 Cell, 50μL pUC19 Control DNA

Linearized, topoisomerase I-activated pcDNA3.1D/V5-His-TOPO vector for 5-minute directional cloning and subsequent high-level expression

  • Proofreading enzyme is used: fewer errors in cloned genes
  • Greater than 90% of clones are in correct orientation for expression, reducing time spent screening colonies for clone orientation
  • CMV promoter: high expression levels
  • C-terminal V5-His fusion tag for easy detection of recombinant protein with an Anti-V5 Antibody
  • 6xHis tag for rapid protein purification on nickel-chelating resin

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type TOPO Expression Kit
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Box 1 (Store at -20°C):
• 20 μl pcDNA™3.1D/V5-His-TOPO™ (15–20 ng/μl)
• 10 μl dNTP Mix
• 50 μl Salt Solution
• 1 ml Sterile Water
• 20 μl T7 Sequencing Primer (0.1 μg/μl)
• 20 μl BGH Reverse Sequencing Primer (0.1 μg/μl)
• 10 μl Control PCR Template (0.1 μg/μl)
• 10 μl Control PCR Primers (0.1 μg/μl each)
• 10 μl Expression Plasmid (0.5 μg/μl)

Box 2 (Store at -80°C):
• 6 ml S.O.C. Medium (may be stored at 4°C or room temperature)
• 21 × 50 μl TOP10 Cells
• 50 μl pUC19 Control DNA
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Directional TOPO
Quantity 20 Reactions
Vector Directional TOPO Vectors, pcDNA
Product Line TOPO, pcDNA
For Use With (Application) Constitutive Expression
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.