Learn More
Invitrogen™ pIB/V5-His Vector Kit
Catalog No. V802001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
V802001 1 kit
1 options
Catalog No. V802001 Supplier Invitrogen™ Supplier No. V802001
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart
Includes
The InsectSelect BSD Kit includes 20μg each of supercoiled, lyophilized pIB/V5-His and pIB/V5-His/CAT, 2μg of lyophilized OpIE2 Forward and Reverse Sequencing primers, 50mg of Blasticidin, Cellfectin Reagent, and 1L of medium for the insect cell line chosen. Sf9 cells are supplied with GIBCO Grace's Insect Media. High Five cells are supplied with GIBCO Express Five Serum-Free Medium. The pIB/V5-His Vector Kit contains vectors and primers.
The pIB/V5-His vector has several features that facilitate expression, analysis, and detection of recombinant protein in insect cells
  • The OpIE2 promoter for constitutive expression
  • The Blasticidin resistance gene for rapid selection of stably transfected cells in two weeks
  • C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Anti-V5 Antibody and rapid purification with nickel-chelating resin
Constitutive Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter OplE2
Product Type Insect Cell Expression Vector
Content And Storage The InsectSelect pIB/V5-His Vector Kit includes 20 μg each of supercoiled lyophilized pIB/V5-His and pIB/V5-CAT vectors, and 2 μg lyophilized OplE2 Forward and Reverse sequencing primers.
Store all reagent at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag Position (to your gene) C-terminal
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pIB
Product Line InsectSelect
For Research Use Only. Not for use in diagnostic procedures.