Learn More
Invitrogen™ pIZT/V5-His Vector Kit
Catalog No. V801001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
V801001 1 kit
1 options
Catalog No. V801001 Supplier Invitrogen™ Supplier No. V801001
Add to Cart
Add to Cart
Includes
The InsectSelect Glow Kit includes 20μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1g Zeocin, Cellfectin Reagent, Sf9 cells, and 1 L of GIBCO Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above.
For high-level expression of the gene of interest
  • The OpIE2 promoter for constitutive expression
  • The Zeocin resistance gene for rapid selection of stably transfected cell lines
  • C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Anti-V5 Antibody and rapid purification with nickel-chelating resin
  • In addition, pIZT/V5-His expresses a fusion of the green fluorescent protein and the Zeocin resistance protein (Zeo-GFP)
  • The Zeo-GFP fusion protein permits rapid selection of stably transfected cell lines with Zeocin and confers a fluorescent phenotype that simplifies identification of transfected cells
Constitutive Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter OplE2
Product Type Insect Cell Expression Vector
Content And Storage The InsectSelect™ Glow Kit includes 20 μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2 μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1 g Zeocin,™ Cellfectin™ Reagent, Sf9 cells, and 1 L of GIBCO™ Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above. Store frozen insect cells in liquid nitrogen. Store medium and Cellfectin™ Reagent at +4°C. Store all other reagents at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag Position (to your gene) C-terminal
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pIZ
Product Line InsectSelect
For Research Use Only. Not for use in diagnostic procedures.