Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plasmid pBR322

Catalog No. 15367014
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-367-014 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-367-014 Supplier Invitrogen™ Supplier No. 15367014
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

pBR322 is supplied at 0.25μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA.

Prepacked column

Plasmid pBR322 (1) contains three unique restriction endonuclease recognition sites within the !-lactamase (AmpR) gene and eight unique sites within the TetR gene. The plasmid, purified from DH10B E. coli, also has 14 non-selectable unique restriction endonuclease recognition sites. Unique cleavage sites for Hind III and Cla I are found in the promoter of the tetracycline resistance gene. Insertion of DNA at either of these two sites usually results in loss of tetracycline resistance. Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis. Transformation of E. coli HB101 to confirm AmpR and TetR is confirmed by plating on selective medium.

Cloning, Restriction Enzyme Cloning

Order Info

Approved for shipment on Wet or Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Restriction Enzyme/ MCS
Concentration 0.25 μg/μl
Content And Storage pBR322 is supplied at 0.25 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C.
For Use With (Application) Cloning
Product Type Plasmid
Quantity 25 μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Vector pBR322
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.