Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ 96 Receiver Plates

Catalog No. 12193025
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-193-025 50 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-193-025 Supplier Invitrogen™ Supplier No. 12193025
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

Includes

The PureLink 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A.

PureLink 96 Plasmid Purification Systems protocol, up to 96 plasmid DNA samples can be simultaneously purified.

  • Centrifugation method eliminates cross-contamination
  • PureLink 96 Plasmid Purification System is designed for high-throughput isolation of high-copy number plasmids
  • PureLink 96 Plasmid Purification Systems protocol, up to 96 plasmid DNA samples can be simultaneously purified
  • Novel technology eliminates time-consuming cell harvesting and alkaline lysis steps, saving time and effort
  • Unique centrifugation-based format of the PureLink 96 Plasmid Purification System eliminates opportunities for cross-contamination between wells, critical factor in PCR and microarrays
  • Isolated plasmid DNA is ideal for use in automated sequencing reactions, offering superior read lengths and data quality
  • Samples may be processed 96 at time or in smaller increments
  • PureLink 96 Plasmid Purification System is compatible with variety of centrifuges, robots, pipetting devices, automated sequencers, and sequencing chemistries
  • PureLink 384 Plasmid Purification Plate is designed for ultra high throughput DNA isolation
  • By utilizing same technology as PureLink 96 Plasmid Purification System, the PureLink 384 Plasmid Purification Plate reduces time and effort while reliably isolating high-quality plasmid DNA for DNA sequencing, PCR, and restriction digestion
  • PureLink 96: Sample size 1.3 mL culture; Typical yields up to 3–5 μg
  • PureLink 384: Sample size 200 μL culture; Typical yields up to 2–3 μg

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The PureLink™ 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A. Components are also available separately. PureLink™ 384 Plasmid Purification Plates are available in packs of 25. PureLink™ 384 Pre-Lysis Buffer is also available separately. Store at room temperature. All components are guaranteed stable for 2 years when properly stored.
Format 96-well Plate
For Use With (Equipment) Robotic Liquid Handling Systems
For Use With (Application) Automated Sequencing Reaction
Plate Type Receiver Plate
Product Line PureLink
Product Type Receiver Plate
Quantity 50 Plates
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.