Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Foil Tape

Catalog No. 12261012
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-261-012 50 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-261-012 Supplier Invitrogen™ Supplier No. 12261012
  • $154.00
    $141.65 / Each of 1

    Save $12.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Package of 50 pieces

PureLink Foil Tape offers a unique approach to high throughput plasmid DNA isolation in the PureLink 96 Plasmid Purification System.

PureLink Foil Tape offers a unique approach to high throughput plasmid DNA isolation in the PureLink 96 Plasmid Purification System.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Content And Storage Package of 50 pieces The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.
For Use With (Application) Automated Nucleic Acid Purification, DNA Extraction
Plate Compatibility 384-well Plates, 96-well Plates
Product Line PureLink
Product Type Foil Tape
Quantity 50 Pieces
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.