Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Growth Blocks

Catalog No. 12256020
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Blocks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-256-020 50 Blocks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-256-020 Supplier Invitrogen™ Supplier No. 12256020
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Package of 50 blocks

PureLink Growth Blocks offer unique approach to high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System.

PureLink Growth Blocks offer unique approach to high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Package of 50 blocks The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.
Format 96 Deep-well Plate
For Use With (Application) Automated Nucleic Acid Purification, DNA Extraction
Product Line PureLink
Product Type Growth Block
Quantity 50 Blocks
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.