Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Viral Lysis Buffer

Catalog No. 12282500
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-282-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-282-500 Supplier Invitrogen™ Supplier No. 12282500
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Filter Plate, Receiver plates, deep-well blocks, foil tape, carrier RNA, proteinase K, Viral Lysis Buffer, Wash Buffer, and RNase-free water.

PureLink 96 Viral RNA/DNA Kit is 96-well silica plate system that can extract both viral RNA and DNA from 200-μL cell-free fluids which uses low-depth 96-well centrifuge bucket for ease-of-use.

  • PureLink 96 Viral RNA/DNA Kit is 96-well silica plate system that can extract both viral RNA and DNA from 200-μL cell-free fluids which uses low-depth 96-well centrifuge bucket for ease-of-use
  • Provide flexibility in centrifuge choice. Minimum speeds for 96 well processing of 2250 x g and rotor bucket depth of 5 cm make this kit one of the most adaptable systems for existing laboratory centrifuges

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Content And Storage The PureLink™ 96 Viral RNA/DNA Kit contains a PureLink™ Filter Plate, Receiver plates, deep-well blocks, foil tape, carrier RNA, proteinase K, Viral Lysis Buffer, Wash Buffer, and RNase-free Water. The kit is shipped and stored at room temperature and the lyophilized tRNA should be removed and stored at -20°C upon receipt. The proteinase K should be stored at +4°C.
For Use With (Application) Nucleic Acid Extraction
Product Line PureLink
Product Type Viral Lysis Buffer
Quantity 500 mL
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.