Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quant-it™ RiboGreen Reagent and RNA Assay Kit

Catalog No. r11490
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 mL
Product Type:
Quant-iT RiboGreen RNA Kit
Quant-iT RiboGreen RNA Reagent
RediPlate 96 RiboGreen RNA Kit
Quantitation Range:
1 to 200 ng
3 to 200 ng
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Quantitation Range
R11490 1 kit Quant-iT RiboGreen RNA Kit 1 to 200 ng
R11491 1 mL Quant-iT RiboGreen RNA Reagent 1 to 200 ng
R32700 1 kit RediPlate 96 RiboGreen RNA Kit 3 to 200 ng
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R11490 Supplier Invitrogen™ Supplier No. R11490
Only null left
Add to Cart
Add to Cart

Quant-iT RiboGreen RNA Reagent is an ultrasensitive fluorescent nucleic acid stain for quantitating RNA in solution. The Quant-iT RiboGreen RNA Assay Kit comes with reagent, dilution buffer, and rRNA standards to provide a linear detection range of 1–200 ng of RNA.

Quant-iT RiboGreen RNA Assay Kit
Containing one of the most sensitive detection dyes for the quantitation of RNA, the Quant-iT RiboGreen RNA Assay Kit and the RediPlate 96 RiboGreen RNA Quantitation Kit offer linear fluorescence detection in the range of 1–200 ng of RNA. Both assay kits contain Quant-iT RiboGreen RNA reagent as well as buffers and RNA standards.

RediPlate 96 RiboGreen RNA Quantitation Kit
The RediPlate 96 RiboGreen RNA Quantitation Kit is a rapid, simple, and highly sensitive fluorescence-based RNA-quantitation assay. The microplate is pre-loaded with RiboGreen reagent, which shows bright green fluorescence upon binding to RNA or DNA. Just add buffer and samples to the microplate wells, wait 10 minutes, and read the fluorescence in any standard fluorescence-based microplate reader. The versatile RediPlate 96 design is ideal for either high-throughput RNA quantitation or the measurement of a small sample numbers.

The RediPlate 96 RiboGreen RNA Quantitation Kit also offers the following benefits:
Automatable—a 10 minute incubation time with no separation steps makes the assay ideal for automated, high-throughput measurements
Precise—the average coefficient of variation for replicate assays is ≤5% using a robotic liquid handling system
Convenient—each plate includes pre-measured RNA standards and is ready for immediate use
Easy to read—the assay can be performed using standard fluorescence-based microplate readers

Applications
The RediPlate 96 RiboGreen RNA Quantitation Kit is ideal for measuring yields of RNA preparations for microarray experiments, reverse transcription PCR (RT-PCR), differential display PCR, northern blot analysis, S1 nuclease assays, RNase protection assays, and cDNA library preparations.

Quant-iT RiboGreen RNA Reagent
The Quant-iT RiboGreen RNA Reagent is one of the most sensitive fluorescent dyes for the quantitation of RNA in solution, with linear fluorescence detection in the range of 1-200 ng of RNA. This reagent is also included with the Quant-iT RiboGreen RNA Assay Kit and comes complete with buffer and RNA standard.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Excitation/Emission 500/525
For Use With (Equipment) Microplate Reader
No. of Reactions 200-2,000 cuvette assays using a 2 mL assay volume
Quantitation Range 1 to 200 ng
Product Line Quant-iT, RIBOGREEN
Product Type Quant-iT RiboGreen RNA Kit
Quantity 1 kit
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.