Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S.N.A.P.™ Gel Purification Kit

Catalog No. K199925
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K199925 25 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K199925 Supplier Invitrogen™ Supplier No. K199925

missing translation for 'validMainframeMsg'

Includes

Each S.N.A.P. Gel Purification Kit includes enough S.N.A.P. columns, collection vials, and reagents for 25 gel purifications.

The S.N.A.P.™ Gel Purification Kit is designed for rapid purification of DNA fragments from agarose gels. The key is unique S.N.A.P. resin, which is provided ready to use in a spin-column format.

The S.N.A.P.™ Gel Purification Kit is designed for rapid purification of DNA fragments from agarose gels. The key is unique S.N.A.P. resin, which is provided ready to use in a spin-column format. The resin efficiently binds nucleic acids up to 20 kb and eliminates organic extractions, and need to perform time-consuming ethanol precipitations.

  • S.N.A.P.™ Gel Purification Kit yields pure DNA in an easy-to-perform, rapid procedure
  • Excise DNA band from your agarose gel
  • Add NaI solution (provided) and melt the agarose at 42–50°C
  • Load sample onto the S.N.A.P.™ spin column
  • Wash away contaminants
  • Elute DNA in water
TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Content And Storage Each S.N.A.P.™ Gel Purification Kit includes enough S.N.A.P.™ columns, collection vials, and reagents for 25 gel purifications. Store all components at room temperature. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
Isolation Technology Spin Column
Sample Type Gel
For Use With (Application) Cloning, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Label or Dye Ethidium Bromide, SYBR Safe
No. of Reactions 25 Preps
Product Line S.N.A.P.
Product Type Gel Purification Kit
Quantity 25 preps
Target Nucleic Acids in Gel Fragments
Show More Show Less
I'm running an agarose gel with TAE or TBE. Can I use your gel extraction kits?

Yes, both TAE and TBE agarose gels are compatible with our gel extraction kits.

What is the difference between Purelink PureLink Quick Gel Extraction System and S.N.A.P. Gel Purification Kit?

The PureLink kit can be used with both TAE and TBE gels, while the S.N.A.P. Gel Purification Kit is recommended for TAE gels only.

Can you use TBE gels with S.N.A.P. gel purification kits?

No, TBE gels contain borate, which interferes with the sodium iodide solution. Therefore, only TAE gels can be used. We recommend the PureLink Quick Gel Extraction kit for both TAE and TBE agarose gels.

What is the smallest fragment that can be purified with S.N.A.P. columns?

S.N.A.P. mini prep columns have been used to purify fragments of 100 bp in gel purification procedures.

What is the resin in the S.N.A.P. kits?

The column contains a silica/glass fiber based resin. However, the exact composition is proprietary.

What is unique about the S.N.A.P. UV-free Gel Purification Kit?

The S.N.A.P. UV-free Gel Purification Kit utilizes crystal violet as a staining reagent, which can be visualized in non-UV light conditions. This can prevent DNA damage that can be caused by the UV light required by normal ethidium bromide staining. DNA fragments recovered using this kit have higher cloning efficiency. In addition, DNA can be visualized under normal light as a thin violet band while the gel is running and excised as soon as bands are sufficiently resolved.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.