Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SequenceRx Enhancer Solution A

Catalog No. 12238010
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-238-010 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-238-010 Supplier Invitrogen™ Supplier No. 12238010
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed to improve length and quality of sequencing data from difficult sequencing templates

The SequenceRx Enhancer System is a panel of seven co-solvents which are added individually to automated fluorescent DNA sequencing reactions. This system is not designed for use on long stretches of poly A.

  • Useful on templates containing regions with secondary structure such as high GC-content, small nucleotide repeats, hairpins, direct repeats, and inverted repeats
  • Requires no change in protocols-the SequenceRx Solution replaces part of water added to reaction mixture
  • Compatible with all sequencing chemistries and platforms
  • Uses one reaction to determine sequence for all four nucleotides (100 reactions equal 100 templates)

Sanger Sequencing, Sanger Sequencing Technology and Accessories, Sequencing

Order Info

Approved for shipment on Wet or Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Guaranteed stable for 6 months when properly stored.
For Use With (Equipment) Sequencing Instruments (Capillary Electrophoresis-Based)
For Use With (Application) Sequencing
No. of Reactions 200
Product Type Enhancer Solution
Quantity 200 Reactions
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.