Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ TOP10F' Chemically Competent E. coli

Catalog No. p-4924890
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
K206001 20 Reactions
K206040 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K206001 Supplier Invitrogen™ Supplier No. K206001
Only null left
Add to Cart
Add to Cart

Provides quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into plasmid vector

The T7 and Sp6 promoters of the pCRII vector allow in vitro transcription of the insert to produce sense or anti-sense products. The TA Cloning Kit Dual Promoter uses the pCRII cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast and convenient: 15-minute, room-temperature ligation
  • Efficient: blue/white screening and >80% clones with correct insert
  • Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free: eliminates any enzymatic modifications of PCR product
  • Streamlined: does not require use of PCR primers that contain restriction sites
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and Sp6 promoters for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

  • How TA Cloning Works\
  • Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to 3' ends of PCR products
  • Linearized vector supplied in this kit has single 3' deoxythymidine (T) residues
  • Allows PCR inserts to ligate efficiently with vector
  • Kit Configurations
  • TA Cloning Kit is offered in a variety of configurations: with One Shot INVF' Chemically Competent E. coli (K2050-01 and K2050-40), One Shot TOP10F' Chemically Competent E. coli (K2060-01 and K2060-40), and without competent cells (K2750-20 and K2750-40) in 20- and 40-reaction kit sizes

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10F ́
Cell Type Chemically Competent E. coli
Cloning Method TA Cloning
Content And Storage This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
For Use With (Application) PCR Cloning
Includes Linearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot TOP10F' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
No. of Reactions 20 Reactions
Product Line One Shot
Product Type Cloning Kit
Promoter T7, SP6
Quantity 20 reactions
Vector pCRII
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.