Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ 0.5M EDTA, pH 8.0

Catalog No. 15575020
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-575-020 4 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-575-020 Supplier Invitrogen™ Supplier No. 15575020
  • $120.00
    $107.65 / Case of 4

    Save $12.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

4 x 100mL bottles UltraPure 0.5M EDTA, pH 8.0.

Sterile-filtered solution

  • Aqueous solution prepared by dissolving Na2EDTA•2H2O in distilled, deionized water and adjusting the pH to 8.0 with sodium hydroxide
  • Suitable for biochemistry or molecular biology applications requiring chelator of divalent metal ions

Agarose Gel Electrophoresis, ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Maxiprep, Midiprep, Miniprep, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification, RNAi, Epigenetics & Non-Coding RNA Research

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
Concentration 0.5 M
Content And Storage Contents: 4 bottles (100 mL each) of UltraPure™ 0.5M EDTA, pH 8.0. Store at room temperature.
For Use With (Application) Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting
pH pH 8.0
Product Line UltraPure
Product Type EDTA
Quantity 4 x 100 mL
Purity Molecular Biology Grade
Shipping Condition Room Temperature
Form Liquid
Show More Show Less
I received a shipment of UltraPure 0.5M EDTA, pH 8.0 (Cat. No. 15575020), but it was stored below 15-30 degrees C over the weekend. Can you confirm this product will still be stable?

We do not have specific data for temperatures below 15 degrees C for UltraPure 0.5M EDTA, pH 8.0 (Cat. No. 15575020). However, as long as the product was not frozen, the performance should not be affected.

What is the difference between EDTA and EGTA?

EDTA binds Mg2+ and Ca2+, while EGTA specifically binds to Ca2+. The use of EGTA may be favored in case where subsequent treatment with Mg2+ dependent enzymes is planned.

How does EDTA work?

EDTA reversibly binds Mg2+, Ca2+ as well as other divalent cations, thus inhibiting enyzmes that require these cations for activity.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.