Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ 1 M Tris-HCI Buffer, pH 7.5
GREENER_CHOICE

Catalog No. 15567027
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-567-027 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-567-027 Supplier Invitrogen™ Supplier No. 15567027
  • $83.75
    $76.65 / Each of 1

    Save $7.10

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

UltraPure 1M Tris-HCl, pH 7.5

Pre-mixed, pH-adjusted, sterile-filtered solution

  • Prepared as 1M concentrate, buffer can be diluted to desired concentration
  • Used in molecular biology or general biochemistry applications
  • Performance and quality testing: No DNase, RNase, or protease activity detected
  • Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 M
Content And Storage Store in refrigerator (2-8°C). UltraPure™ 1M Tris-HCl, pH 7.5 is guaranteed for 6 months from date of purchase unless otherwise indicated in product documentation.
pH pH 7.5
Product Line UltraPure
Product Type Tris-HCl Buffer
Quantity 1 L
Shipping Condition Room Temperature
Form Liquid
How should I store UltraPure 1 M Tris-HCI Buffer, pH 7.5 (Cat. No. 15567027)?

We recommend storing UltraPure 1 M Tris-HCI Buffer, pH 7.5 (Cat. No. 15567027) in the refrigerator (2-8 degrees C).

Do you offer any alternatives for UltraPure 1M Tris-HCl Buffer, pH 7.5 (Cat. No. 15567027)?
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.