Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Calf Thymus DNA Solution

Catalog No. 15633019
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-633-019 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-633-019 Supplier Invitrogen™ Supplier No. 15633019
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blocks non-specific attachment of probe to surface of a membrane

UltraPure DNA solutions from Calf Thymus is ready-to-use, sheared DNA solutions that is used directly in the preparation of prehybridization and hybridization solutions.

  • DNA solution prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water and are sheared to an average size of ≤2,000bp
  • Concentration is adjusted to 10mg/mL
  • Performance and Quality Testing: Concentration, size verification, viscosity, and ultraviolet absorbance (A260/A280) are analytically tested

ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, Epigenetics & Non-Coding RNA Research, Southern Blotting

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
For Use With (Application) Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting
Key Functions Block Non-Specific Hybridization
Product Line UltraPure
Product Type Calf Thymus DNA Solution
Quantity 5 x 1 mL
Reagent Type Calf Thymus DNA
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.