Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Denhardt's Solution (50X)

Catalog No. 750018
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-001-8 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-001-8 Supplier Invitrogen™ Supplier No. 750018
  • $245.00
    $220.65 / Each of 1

    Save $24.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile filtered

For use with: 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion Personal Genome Machine™ (PGM™) System, KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMA

  • Mixture of blocking agents used in membrane-based hybridization protocols
  • Contains 1% Ficoll (type 400), 1% polyvinylpyrrolidone, and 1% bovine serum albumin

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Blotting Buffer
Content And Storage Store in freezer (-5 to -30°C).
Product Line UltraPure
Product Type Denhardt's Solution
Quantity 100 mL
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.