Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Dithiothreitol

Catalog No. 15508013
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-508-013 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-508-013 Supplier Invitrogen™ Supplier No. 15508013
  • $199.00
    $186.65 / Each of 1

    Save $12.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure Dithiothreitol (DTT, Cleland's reagent), which reduces disulfides to their corresponding thiols, is used at low concentrations to stabilize enzymes containing free sulfhydryl groups.

UltraPure Dithiothreitol (DTT, Cleland's reagent), which reduces disulfides to their corresponding thiols, is used at low concentrations to stabilize enzymes containing free sulfhydryl groups. Higher concentrations of DTT are used to cleave disulfide linkages in polypeptides and to facilitate protein denaturation by detergents or chaotropic agents.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C).
For Use With (Application) Protein Electrophoresis & Western Blotting
Product Line UltraPure
Quantity 5 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Can beta-mercaptoethanol (BME) be used rather than DTT as the reducing agent in the NuPAGE LDS Sample Buffer?

Either BME or DTT can be used in the NuPAGE LDS Sample Buffer.

Make sure that a fresh solution of BME is used. FINAL concentration:

DTT 50-100 mM

BME 2-5%

How do I prepare a 1 M DTT solution from powder and how should I store the solution?

Preparing 1 M DTT:
1) Dissolve 3.09 g DTT in 20 mL distilled H2O.
2) Dispense into 1 mL aliquots.
3) Store the aliquots at -20 degrees C.
Note: Upon thawing the vial, there may be some precipitation that can be re-dissolved by warming and mixing.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.