Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O)

Catalog No. 15576028
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-576-028 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-576-028 Supplier Invitrogen™ Supplier No. 15576028
  • $176.00
    $165.65 / Each of 1

    Save $10.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for biochemistry or molecular biology applications

  • UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA•2H2O), is a chelator of divalent metal cation

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
Content And Storage Store at room temperature.
For Use With (Application) Biochemistry, Molecular Biology
Product Line UltraPure
Product Type Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate
Quantity 500 g
Purity Molecular Biology Grade
Shipping Condition Room Temperature
Form Powder
Can I use EDTA (Cat. No. 17892) in molecular biology? If not, can you provide an alternative?

EDTA (Cat. No. 17892) is not intended for molecular biology applications.

UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O) (Cat. No. 15576028) is a potential alternative intended for molecular biology applications.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.