Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Formamide

Catalog No. 15515026
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-515-026 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-515-026 Supplier Invitrogen™ Supplier No. 15515026
 Restricted Item
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure Formamide is commonly used for denaturing nucleic acids for sequencing gel electrophoresis, electron microscopy, and hybridization.

UltraPure Formamide is commonly used for denaturing nucleic acids for sequencing gel electrophoresis, electron microscopy, and hybridization. A liquid at room temperature, UltraPure Formamide is vacuum distilled and packaged under dry nitrogen and can be used as supplied for many applications within three months of date of purchase. Because the breakdown products of formamide degrade nucleic acids, for sensitive applications formamide should be deionized by treatment with a mixed-bed ion-exchange resin immediately before use.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
For Use With (Application) Cellular Imaging/Nucleic Acid Gel Electrophoresis & Blotting
Product Type Formamide
Product Line UltraPure
Purity DNase-Free, RNase-Free
Quantity 500 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Liquid
Can a sample buffer with formamide be used with the Invitrogen TBE-Urea system?

There are many sample buffer formulations used, however we have found a distinct difference in the band appearance depending on the sample buffer composition. After evaluating urea, formamide, and various buffer systems, we found that the sharpest, flattest bands were obtained with a urea, Ficoll, and TBE buffer solution. Sample buffers made with formamide resulted in fuzzy, indistinct bands.

Is formamide (Cat. No. 15515-026) supplied as a liquid? When is deionization required, and can you provide a protocol?

Yes, formamide is supplied as a liquid even though it is sold by weight.

It is appropriate to use out of the bottle for most applications (hybridization buffer, loading buffers for sequencing gels, and formaldehyde-containing RNA gels) if less than one year old and stored properly (-20 degrees C). (If formamide smells like ammonia, it is probably breaking down and should not be used.) However, we have seen that in a protocol we have for running RNA gels without formaldehyde in the gel, freshly deionized formamide is essential for preparation of the loading buffer. That protocol as well as the protocol for deionizing the formamide follows:

These reagents are used in electrophoresis of RNA in agarose gels in 1X MOPS EDTA buffer. (Note: no formaldehyde is used in the gel.) The sample is prepared in the sample buffer (1.5 to 3 µL sample mixed with sample buffer). Heat the RNA in sample buffer for 10 min at 65 degrees C. Place on ice immediately then load gel.

10X MOPS EDTA (10X ME)

(1) Measure out 700 mL DEPC-treated water in a DEPC-treated 1-liter graduated cylinder and place in a DEPC-treated 1-liter beaker.
(2) Place beaker on stir plate and begin stirring with a DEPC-treated stir bar.
(3) Add 104.65 g MOPS to beaker. Weigh out MOPS using a sterile-flamed spatula from an unopened bottle MOPS or one set aside for RNA only. Dissolve by stirring.
(4) Add 40 mL 0.25 M EDTA to solution. Note: the EDTA solution should be made with DEPC-treated processed water.
(5) Adjust pH to 7.0 by adding 10 N NaOH (~20 mL)
(6) Bring up to 1 liter volume with DEPC-treated water.
(7) Filter solution using a 0.2 µm Nalgene filter unit.
(8) Store at 4 degrees C for six months in a DEPC-treated glass bottle. (This prevents solution from yellowing.) NOTE: The presence of yellow color does not affect the performance of the buffer.

Formamide Deionization protocol:
1. Add 1 g mixed bed, ion exchange resin for every 10 ml of formamide. Suitable ion exchange resins include BioRad AG501-X8 and Fisher Resin 1-300.
2. Stir for 30 to 60 min @ RT.
3. Filter through Whatman No. 1 filter paper.
4. Dispense into units of use and store at -20°C

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.