Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Guanidine Hydrochloride

Catalog No. 15502016
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-502-016 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-502-016 Supplier Invitrogen™ Supplier No. 15502016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6M) guanidine hydrochloride solution, many proteins unfold and separate into their constituent polypeptide chains.

UltraPure Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6M) guanidine hydrochloride solution, many proteins unfold and separate into their constituent polypeptide chains. Guanidine hydrochloride is widely used to purify nucleic acids from cell extracts and is useful as an RNase inhibitor during the isolation of mRNA from mammalian cells.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) RNA Extraction
Packaging Type Bottle
Product Type Guanidine Hydrochloride
Product Line UltraPure
Purity DNase-Free, RNase-Free
Quantity 500 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.