Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ N,N'-Methylenebisacrylamide

Catalog No. 15516024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-516-024 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-516-024 Supplier Invitrogen™ Supplier No. 15516024
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polymerization time is evaluated in SDS polyacrylamide gel system

  • UltraPure N,N′-Methylenebisacrylamide is most frequently used cross-linking agent for polyacrylamide gels

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Gel Casting, Nucleic Acid Gel Electrophoresis & Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Gel Electrophoresis
Product Line UltraPure
Product Type N,N'-Methylenebisacrylamide
Quantity 100 g
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.