Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Sucrose

Catalog No. 15503022
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-503-022 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-503-022 Supplier Invitrogen™ Supplier No. 15503022
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure Sucrose is suitable for forming density gradients for a variety of separation applications.

UltraPure Sucrose is suitable for forming density gradients for a variety of separation applications. It is used to separate RNAs, to prepare bacteriophage λ-DNA arms, and to separate proteins. UltraPure Sucrose gradients typically are formed by use of gradient formers or by layering two or more sucrose solutions in varying amounts in a centrifuge tube.

Performance and Quality Testing
No protease activity detected.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sucrose
Content And Storage Store at room temperature.
For Use With (Application) Density Gradients - Separation Applications
Product Line UltraPure
Purity Molecular Biology Grade
Quantity 5 kg
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.