Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Tris Hydrochloride

Catalog No. 15506017
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-506-017 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-506-017 Supplier Invitrogen™ Supplier No. 15506017
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Simplifies preparation of Tris buffers

  • UltraPure Tris Hydrochloride [tris(hydroxymethyl)aminomethane hydrochloride] used for simplify preparation of Tris buffers

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris
Content And Storage Store at room temperature.
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type Tris Hydrochloride
Quantity 500 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.