Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Agarose-1000
GREENER_CHOICE

Catalog No. 16550100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-550-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-550-100 Supplier Invitrogen™ Supplier No. 16550100
  • $506.00
    $465.65 / Each of 1

    Save $40.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

100g pouch

Easy-pour spout reduces spills and contamination

UltraPure Agarose-1000 is polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis. UltraPure Agarose 1000 is specifically formulated for the high-resolution separation of small (<1,000 bp) DNA, RNA, and PCR fragments. This standard melting temperature agarose can resolve DNA fragments differing in as little as 10bp in size

  • Ideal for analysis and recovery of small DNA and RNA fragments
  • Strong gel structure allows for better handling
  • Improved clarity allows for enhanced visibility of fragments
  • Improved packaging: Environmentally friendly, sustainable packaging uses 75% less plastic than the original bottles
  • Performance and quality testing: Performance of UltraPure Agarose 1000 is evaluated to satisfy specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase assay Separation range: 50bp to 1000bp

UltraPure Agarose 10000 (Cat. No. 16550-100) is a replacement for Cat. No. 10975-035.

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 pouch containing 100 g of UltraPure™ Low Melting Point Agarose. Store at room temperature.
Separation Range 50 to 1000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Melting Point Standard Melting Point
Quantity 100 g
Shipping Condition Room Temperature
Product Line UltraPure
Product Type Agarose Gel
I want to pour my own gels. Which agarose should I use?

Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.