Learn More
Bio IVT INVITROGRO CP Medium, 500 mL, plating medium, contains serum, without antibiotics, optimized for cryoplateable hepatocytes
Catalog No. NC2201318
Encompass
INVITROGRO CP Medium, 500 mL, plating medium, contains serum, without antibiotics, optimized for cryoplateable hepatocytes
This item is not returnable. View return policy
Catalog No. NC2201318 Supplier Bio IVT Supplier No. Z990003
INVITROGRO CP Medium, 500 mL, plating medium, contains serum, without antibiotics, optimized for cryoplateable hepatocytes