Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Invitrosol™ LC/MS Protein Solubilizer Kit

Catalog No. MS10007
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS10007 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS10007 Supplier Invitrogen™ Supplier No. MS10007
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrosol™ LC/MS Protein Solubilizer Kit

The Invitrosol™ LC/MS (Liquid Chromatorgraphy/Mass spectrometry) Protein Solubilizer Kit contains a novel, proprietary surfactant blend for simplified preparation of hydrophobic and hydrophilic proteins or peptides for ESI (Electrospray Ionization) mass spectrometry (MS) experiments.

Optimized for LC/MS analysis, Invitrosol LC/MS Protein Solubilizer greatly improves data quality by removing detergents that are incompatible with MS ionization techniques (e.g. CHAPs, Triton-X, SDS, etc.). Invitrosol™ LC/MS Protein Solubilizer is compatible with enzymatic proteolysis and improves statistical confidence in mass spectral results for hydrophobic proteins and peptides. Additionally, Invitrosol™ LC/MS Protein Solubilizer does not interfere with your chromatography separations (Figure 1) and does not contaminate LC/MS instrumentation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Protein Solubilizer Kit
Product Line Invitrosol
Content And Storage The Invitrosol™ LC/MS Protein Solubilizer Kit contains surfactant blend and standard BSA tryptic digest. Store at -20°C. Guaranteed stable for 6 months if properly stored.
Workflow Step Protein Sample Preparation
Quantity 1 kit
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.