Learn More
Bio X Cell InVivoMAb, Anti-Human MHC I, W6/32, 25 MG
Supplier: Bio X Cell BE0079R025MG
The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, and C. This antibody plays a central role in cell-mediated immune responses and tumor surveillance.
- Target human MHC Class I (HLA-A, HLA-B, HLA-C)
- Clone W6/32
- Mouse IgG2a, κ isotype
- Purity is ≥95% determined by SDS-PAGE
- Endotoxin is ≤1EU/mg (≤0.001EU/μg)
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Purified from cell culture supernatant in an animal-free facility
- Protein G purification
- Sterility is 0.2 μm filtration
- Suitable for functional assays, *in vitro* Organoids/Organ-on-Chip, immunopeptidomics, immunoprecipitation, immunofluorescence, flow cytometry, and ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.