Learn More
Bio X Cell InVivoPlus Mouse Isotype Control, IgG2a, 100 MG
Supplier:  Bio X Cell BP0085R100MG
Encompass_Preferred
The InVivoPlus mouse IgG2a isotype control is an antibody solution with unknown specificity, provided in a 100 mg quantity. It serves as a crucial control for in vivo antibody experiments.
  • Mouse host
  • IgG2a, κ isotype
  • Store at 4°C; do not freeze
Catalog No. NC2095509
Add to Cart
Request a Quote