Learn More
Bio X Cell InVivoMAb Anti-Canine PD-L1 (B7-H1) Monoclonal Antibody, Clone JC071, 5 MG
Supplier: Bio X Cell BE0390R005MG
Bio X Cell InVivoMAb anti-canine PD-L1 (B7-H1) (clone JC071) is a mouse monoclonal antibody that recognizes canine PD-L1 (CD274/B7-H1) and is suitable for detection and functional assays in canine samples. The antibody is supplied in multiple sizes, formulated in PBS, and should be stored at 4C without freezing.
- Reacts with canine PD-L1 (B7-H1/CD274).
- Clone JC071.
- Host species: mouse; isotype: IgG1, .
- Suitable for detection and functional assays, including flow cytometry and Western blot.
- Formulation: PBS, pH 7.0, no stabilizers or preservatives.
- Storage: store at 4C; do not freeze.
- Available sizes: 1 mg, 5 mg, 25 mg, 50 mg, 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.