Learn More
BIOHIPPO InVivoMAb Anti-HSV1/HHV1 gD/Envelope glycoprotein D (Fd138-80), 100 µg
SDP
Catalog No. 5000860572
Encompass_Preferred
HSV1/HHV1 gD/Envelope glycoprotein D Recombinant Antibody (clone Fd138-80) is a recombinant Human IgG1 antibody specific for HSV1/HHV1 gD/Envelope glycoprotein D. It reacts with Human herpesvirus 1. Suitable for Blocking, ELISA, FCM. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60572 Supplier BIOHIPPO Supplier No. BHA21409736
HSV1/HHV1 gD/Envelope glycoprotein D Recombinant Antibody (clone Fd138-80) is a recombinant Human IgG1 antibody specific for HSV1/HHV1 gD/Envelope glycoprotein D. It reacts with Human herpesvirus 1. Suitable for Blocking, ELISA, FCM. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.