Learn More
Bio X Cell InVivoMAb Anti-Human NKG2D (CD314), Purified Mouse Monoclonal Antibody, Clone 1D11, 1 mg
Supplier: Bio X Cell BE0351R001MG/DEL
A purified mouse monoclonal antibody (clone 1D11) specific for human NKG2D (CD314), supplied as a 1 mg vial in PBS, pH 7.0. The antibody is intended for in vitro NKG2D blocking, immunoprecipitation, and flow cytometry, with low endotoxin and refrigerated storage requirements.
- Purified mouse monoclonal antibody, clone 1D11.
- Specific for human NKG2D (CD314).
- Supplied at 1 mg vial for in vitro use.
- Validated for blocking, immunoprecipitation, and flow cytometry.
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Endotoxin ≤1 EU/mg as determined by LAL assay.
- Store at 4°C at stock concentration; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.