Learn More
BIOHIPPO InVivoMAb Anti-Human CD340/ERBB2/HER2/NEU Antibody (520C9), 100 µg
SDP
Catalog No. 5000860059
Encompass_Preferred
CD340/ERBB2/HER2/NEU Recombinant Antibody (clone 520C9) is a recombinant Mouse IgG2a, kappa antibody specific for CD340/ERBB2/HER2/NEU. It reacts with Human. Suitable for Blocking, ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60059 Supplier BIOHIPPO Supplier No. BHA21409735
CD340/ERBB2/HER2/NEU Recombinant Antibody (clone 520C9) is a recombinant Mouse IgG2a, kappa antibody specific for CD340/ERBB2/HER2/NEU. It reacts with Human. Suitable for Blocking, ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.