Learn More
BIOHIPPO InVivoMAb Anti-Human CSF1/M-CSF (Iv0052), 100 µg
SDP
Catalog No. 5000860369
Encompass_Preferred
CSF1/M-CSF Recombinant Antibody (clone Iv0052) is a recombinant Human IgG1, kappa antibody specific for CSF1/M-CSF. It reacts with Human. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60369 Supplier BIOHIPPO Supplier No. BHA21409944
CSF1/M-CSF Recombinant Antibody (clone Iv0052) is a recombinant Human IgG1, kappa antibody specific for CSF1/M-CSF. It reacts with Human. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.