Learn More
BIOHIPPO InVivoMAb Anti-Human TGFB1/TGF-beta-1 (Iv0069), 100 µg
SDP
Catalog No. 5000859494
Encompass_Preferred
TGFB1/TGF-beta-1 Recombinant Antibody (clone Iv0069) is a recombinant Human IgG4, kappa antibody specific for TGFB1/TGF-beta-1. It reacts with Human. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-59494 Supplier BIOHIPPO Supplier No. BHA21409927
TGFB1/TGF-beta-1 Recombinant Antibody (clone Iv0069) is a recombinant Human IgG4, kappa antibody specific for TGFB1/TGF-beta-1. It reacts with Human. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.