Learn More
BIOHIPPO InVivoMAb Anti-Influenza A virus Neuraminidase/NA Antibody (1G01), 100 µg
SDP
Catalog No. 5000859778
Encompass_Preferred
Influenza A virus Neuraminidase/NA Recombinant Antibody (clone 1G01) is a recombinant Human IgG1 antibody specific for Influenza A virus Neuraminidase/NA. It reacts with Influenza A virus. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-59778 Supplier BIOHIPPO Supplier No. BHA21409815
Influenza A virus Neuraminidase/NA Recombinant Antibody (clone 1G01) is a recombinant Human IgG1 antibody specific for Influenza A virus Neuraminidase/NA. It reacts with Influenza A virus. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.