Learn More
BIOHIPPO InVivoMAb Anti-MERS-CoV S2/Spike glycoprotein 2 Antibody (1.6C7), 100 µg
SDP
Catalog No. 5000860198
Encompass_Preferred
MERS-CoV S2/Spike glycoprotein 2 Recombinant Antibody (clone 1.6C7) is a recombinant Mouse IgG2a, kappa antibody specific for MERS-CoV S2/Spike glycoprotein 2. It reacts with Middle East respiratory syndrome-related coronavirus. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60198 Supplier BIOHIPPO Supplier No. BHA21409700
MERS-CoV S2/Spike glycoprotein 2 Recombinant Antibody (clone 1.6C7) is a recombinant Mouse IgG2a, kappa antibody specific for MERS-CoV S2/Spike glycoprotein 2. It reacts with Middle East respiratory syndrome-related coronavirus. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.