Learn More
Bio X Cell InVivoMAb, Anti-Mouse IL-6R, Clone 15A7, 50 MG
Supplier: Bio X Cell BE0047R050MG
This InVivoMAb anti-mouse IL-6R is a monoclonal rat antibody designed for effective blocking of IL-6/IL-6R signaling in both in vivo and in vitro applications.
- Applications: In vivo and in vitro blocking of IL-6/IL-6R signaling
- Antibody isotype: IgG2b, κ
- Immunogen: OKT-4 hybridoma cells
- Host: Rat
- Clonality: Monoclonal (15A7)
- Conjugate: Unconjugated
- Target: Interleukin-6 receptor subunit alpha (Mouse)
- Reactivity: Mus musculus (House mouse), Homo sapiens (Human)
- Dilutions for FC/FACS: 1:200
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.