Learn More
Bio X Cell InVivoMAb CD19, 1D3, IgG2a, Purified, 1 MG
Supplier: Bio X Cell BE0150R001MG
The 1D3 monoclonal antibody specifically recognizes mouse CD19, a 95 kDa transmembrane glycoprotein critical for B-cell receptor signaling. This antibody is crucial for understanding and targeting lymphoproliferative disorders due to CD19's high expression in many lymphomas and leukemias.
- Clone 1D3 targets mouse CD19
- Host is rat with an IgG2a, κ isotype
- Immunogen is K562 cells expressing extracellular domain of mouse CD19
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1EU/mg (≤0.001EU/μg) by LAL assay
- Purity ≥95% by SDS-PAGE
- Sterility is 0.2 μm filtered
- Produced from cell culture supernatant in an animal-free facility
- Purified by Protein G
- Molecular weight is 150 kDa
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.